Anti-PROKR2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PROKR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-517
- 77795-516
- Antigen symbol:
- PROKR2
- PROKR2
- Antigen name:
- Prokineticin receptor 2 protein (1-45AA)
- Prokineticin receptor 2 protein (1-45AA)
- Antigen synonyms:
- B830005M06Rik|dJ680N4.3|EG-VEGRF2|G protein coupled receptor 73 like 1|G protein coupled receptor I5E|G-protein coupled receptor 73-like 1|GPR73b|GPR73L1|GPRg2|HH3|KAL3|PK R2|PK-R2|PKR2|PKR2_HUMAN|Prokineticin receptor 2
- B830005M06Rik|dJ680N4.3|EG-VEGRF2|G protein coupled receptor 73 like 1|G protein coupled receptor I5E|G-protein coupled receptor 73-like 1|GPR73b|GPR73L1|GPRg2|HH3|KAL3|PK R2|PK-R2|PKR2|PKR2_HUMAN|Prokineticin receptor 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NFJ6
- Q8NFJ6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together