Anti-VAMP4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: VAMP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77804-222
- 77804-223
- Antigen symbol:
- VAMP4
- VAMP4
- Antigen name:
- Vesicle-associated membrane protein 4 protein (1-115AA)
- Vesicle-associated membrane protein 4 protein (1-115AA)
- Antigen synonyms:
- VAMP 24|VAMP 4|VAMP-4|VAMP24|VAMP4 protein|VAMP4_HUMAN|Vesicle associated membrane protein 4|Vesicle associated membrane protein 4 isoform CRA a|Vesicle associated membrane protein 4 isoform CRA b|Vesicle-associated membrane protein 4
- VAMP 24|VAMP 4|VAMP-4|VAMP24|VAMP4 protein|VAMP4_HUMAN|Vesicle associated membrane protein 4|Vesicle associated membrane protein 4 isoform CRA a|Vesicle associated membrane protein 4 isoform CRA b|Vesicle-associated membrane protein 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75379
- O75379
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together