Anti-RBM12 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RBM12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77836-516
- 77836-517
- Antigen symbol:
- RBM12
- RBM12
- Antigen name:
- RNA-binding protein 12 protein (1-200AA)
- RNA-binding protein 12 protein (1-200AA)
- Antigen synonyms:
- HRIHFB2091|KIAA0765|RBM 12|RBM12_HUMAN|RNA binding motif protein 12|RNA binding protein 12|RNA-binding motif protein 12|RNA-binding protein 12|SH3/WW domain anchor protein in the nucleus|SWAN
- HRIHFB2091|KIAA0765|RBM 12|RBM12_HUMAN|RNA binding motif protein 12|RNA binding protein 12|RNA-binding motif protein 12|RNA-binding protein 12|SH3/WW domain anchor protein in the nucleus|SWAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NTZ6
- Q9NTZ6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together