Anti-SRGAP3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: SRGAP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-975
- 77800-974
- Antigen symbol:
- SRGAP3
- SRGAP3
- Antigen name:
- SLIT-ROBO Rho GTPase-activating protein 3 protein (709-955AA)
- SLIT-ROBO Rho GTPase-activating protein 3 protein (709-955AA)
- Antigen synonyms:
- ARHGAP 14|ARHGAP14|Gbi|ME GAP|MEGAP|Mental disorder associated GAP|Mental disorder-associated GAP|Rho GTPase activating protein 14|Rho GTPase-activating protein 14|SLIT ROBO Rho GTPase activating protein 3|SLIT-ROBO Rho GTPase-activating protein 3|srGAP 2|srGAP 3|srGAP2|srGAP3|SRGP 2|SRGP2|SRGP3_HUMAN|WAVE associated Rac GTPase activating protein|WAVE-associated Rac GTPase-activating protein|WRP
- ARHGAP 14|ARHGAP14|Gbi|ME GAP|MEGAP|Mental disorder associated GAP|Mental disorder-associated GAP|Rho GTPase activating protein 14|Rho GTPase-activating protein 14|SLIT ROBO Rho GTPase activating protein 3|SLIT-ROBO Rho GTPase-activating protein 3|srGAP 2|srGAP 3|srGAP2|srGAP3|SRGP 2|SRGP2|SRGP3_HUMAN|WAVE associated Rac GTPase activating protein|WAVE-associated Rac GTPase-activating protein|WRP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43295
- O43295
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together