Anti-TARS2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TARS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-866
- 77801-865
- Antigen symbol:
- TARS2
- TARS2
- Antigen name:
- Threonine--tRNA ligase, mitochondrial protein (369-718AA)
- Threonine--tRNA ligase, mitochondrial protein (369-718AA)
- Antigen synonyms:
- TARS2|TARSL1|Threonine--tRNA ligase|mitochondrial|EC 6.1.1.3|Threonyl-tRNA synthetase|ThrRS|Threonyl-tRNA synthetase-like 1
- TARS2|TARSL1|Threonine--tRNA ligase|mitochondrial|EC 6.1.1.3|Threonyl-tRNA synthetase|ThrRS|Threonyl-tRNA synthetase-like 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BW92
- Q9BW92
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together