Anti-RASSF2 Rabbit Recombinant Polyclonal Antibody
Catalogue # 77796-720
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen name:Ras association domain-containing protein 2 protein (69-326AA)
- Antigen symbol:RASSF2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P50749
- Antigen synonyms:CENP 34|Centromere protein 34|Ras association (RalGDS/AF 6) domain family 2|Ras association (RalGDS/AF-6) domain family member 2|Ras association domain containing protein 2|Ras association domain-containing protein 2|RASF2_HUMAN|RASFADIN|RASSF2|RP4-599I11.1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB: 1:1000 to 1:5000, IP:1:200 to 1:2000
Type:
Antigen: RASSF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human