Anti-SLA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-843
- 77821-842
- Antigen symbol:
- SLA
- SLA
- Antigen name:
- Src-like-adapter protein (2-276AA)
- Src-like-adapter protein (2-276AA)
- Antigen synonyms:
- hSLAP|SLA 1|sla|SLA1|SLAP 1|SLAP|SLAP-1|SLAP1|SLAP1_HUMAN|Src like adapter|Src like adapter protein 1|Src like adaptor|SRC-like-adapter|Src-like-adapter protein 1
- hSLAP|SLA 1|sla|SLA1|SLAP 1|SLAP|SLAP-1|SLAP1|SLAP1_HUMAN|Src like adapter|Src like adapter protein 1|Src like adaptor|SRC-like-adapter|Src-like-adapter protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13239
- Q13239
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together