Anti-TCOF1 Rabbit Polyclonal Antibody
77808-250
New Product
- Antigen name:Synthesized peptide derived from the N-terminal region of Treacle.
- Antigen symbol:TCOF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13428
- Antigen synonyms:Mandibulofacial dysostosis|MFD1|Nucleolar trafficking phosphoprotein|TCOF 1|TCOF_HUMAN|TCOF1|TCS|TCS1|Treacher Collins Franceschetti syndrome 1|Treacher Collins syndrome|Treacher Collins syndrome protein|Treacle|Treacle protein
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA:1:10000
Type:
Antigen: TCOF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse