Anti-MED26 Rabbit Polyclonal Antibody
77812-387
New Product
- Antigen name:Synthesized peptide derived from the N-terminal region of CRSP70.
- Antigen symbol:MED26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O95402
- Antigen synonyms:Activator recruited cofactor 70 kDa component|Activator-recruited cofactor 70 kDa component|ARC 70|ARC70|Cofactor required for Sp1 transcriptional activation subunit 7|Cofactor required for Sp1 transcriptional activation, subunit 7 (70kD)|Cofactor required for Sp1 transcriptional activation, subunit 7, 70kDa|CRSP 7|CRSP complex subunit 7|CRSP70|MED 26|Med26|MED26_HUMAN|Mediator complex subunit 26|Mediator of RNA polymerase II transcription subunit 26|Transcriptional coactivator CRSP70
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:20000
Type:
Antigen: MED26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse