Anti-ompC Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ompC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Specifications
- Catalog No:
- 77817-040
- 77817-041
- Antigen symbol:
- ompC
- ompC
- Antigen name:
- Recombinant Escherichia coli Outer membrane protein C protein (22-367AA)
- Recombinant Escherichia coli Outer membrane protein C protein (22-367AA)
- Antigen synonyms:
- ompC|meoA|par|b2215|JW2203|Outer membrane porin C|Outer membrane protein 1B|Outer membrane protein C|Porin OmpC
- ompC|meoA|par|b2215|JW2203|Outer membrane porin C|Outer membrane protein 1B|Outer membrane protein C|Porin OmpC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06996
- P06996
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together