Anti-BTN2A1 Rabbit Recombinant Antibody (Biotinylated)
New Product
Type:
Antigen: BTN2A1
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-778
- 77830-779
- Antigen symbol:
- BTN2A1
- BTN2A1
- Antigen name:
- Butyrophilin subfamily 2 member A1 protein (29-248AA)
- Butyrophilin subfamily 2 member A1 protein (29-248AA)
- Antigen synonyms:
- BK14H9.1|BT2.1|BT2A1_HUMAN|BTF1|BTN2A1|Butyrophilin BTF1|Butyrophilin subfamily 2 member A1|CTA-14H9.1|DJ3E1.1
- BK14H9.1|BT2.1|BT2A1_HUMAN|BTF1|BTN2A1|Butyrophilin BTF1|Butyrophilin subfamily 2 member A1|CTA-14H9.1|DJ3E1.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7KYR7
- Q7KYR7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together