Anti-PBX1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB:1:500 to 1:5000, IHC:1:100 to 1:300, IF:1:50 to 1:200
Type:
Antigen: PBX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
- Catalog No:
- 77793-305
- 77793-306
- Antigen symbol:
- PBX1
- PBX1
- Antigen name:
- Pre-B-cell leukemia transcription factor 1 protein (315-430AA)
- Pre-B-cell leukemia transcription factor 1 protein (315-430AA)
- Antigen synonyms:
- DKFZp686B09108|Homeo box protein PBX1|Homeo box protein PRL|Homeobox protein PBX 1|Homeobox protein PBX1|Homeobox protein PRL|MGC126627|PBX 1|Pbx1|PBX1_HUMAN|Pre B cell leukemia homeobox 1|Pre B cell leukemia transcription factor 1|Pre-B-cell leukemia transcription factor 1|PRL
- DKFZp686B09108|Homeo box protein PBX1|Homeo box protein PRL|Homeobox protein PBX 1|Homeobox protein PBX1|Homeobox protein PRL|MGC126627|PBX 1|Pbx1|PBX1_HUMAN|Pre B cell leukemia homeobox 1|Pre B cell leukemia transcription factor 1|Pre-B-cell leukemia transcription factor 1|PRL
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P40424
- P40424
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together