Anti-MEF2D Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MEF2D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-058
- 77822-057
- Antigen symbol:
- MEF2D
- MEF2D
- Antigen name:
- Myocyte-specific enhancer factor 2D protein (299-509AA)
- Myocyte-specific enhancer factor 2D protein (299-509AA)
- Antigen synonyms:
- DKFZp686I1536|MADS box transcription factor 2 polypeptide D|Mef2d|MEF2D_HUMAN|Myocyte enhancer factor 2D|Myocyte specific enhancer factor 2, polypeptide D|Myocyte specific enhancer factor 2D|Myocyte-specific enhancer factor 2D
- DKFZp686I1536|MADS box transcription factor 2 polypeptide D|Mef2d|MEF2D_HUMAN|Myocyte enhancer factor 2D|Myocyte specific enhancer factor 2, polypeptide D|Myocyte specific enhancer factor 2D|Myocyte-specific enhancer factor 2D
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14814
- Q14814
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together