Anti-SLC25A23 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC25A23
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-009
- 77838-010
- Antigen symbol:
- SLC25A23
- SLC25A23
- Antigen name:
- Calcium-binding mitochondrial carrier protein SCaMC-3 protein (1-188AA)
- Calcium-binding mitochondrial carrier protein SCaMC-3 protein (1-188AA)
- Antigen synonyms:
- SLC25A23|APC2|MCSC2|SCAMC3Calcium-binding mitochondrial carrier protein SCaMC-3|Mitochondrial ATP-Mg/Pi carrier protein 2|Mitochondrial Ca(2+)-dependent solute carrier protein 2|Small calcium-binding mitochondrial carrier protein 3|Solute carrier family 25 member 23
- SLC25A23|APC2|MCSC2|SCAMC3Calcium-binding mitochondrial carrier protein SCaMC-3|Mitochondrial ATP-Mg/Pi carrier protein 2|Mitochondrial Ca(2+)-dependent solute carrier protein 2|Small calcium-binding mitochondrial carrier protein 3|Solute carrier family 25 member 23
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BV35
- Q9BV35
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together