Anti-NBEAL2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NBEAL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-469
- 77830-470
- Antigen symbol:
- NBEAL2
- NBEAL2
- Antigen name:
- Neurobeachin-like protein 2 protein (1360-1574AA)
- Neurobeachin-like protein 2 protein (1360-1574AA)
- Antigen synonyms:
- BDPLT4|GPS|NBEAL2|NBEL2_HUMAN|Neurobeachin-like protein 2|UNQ253/PRO290
- BDPLT4|GPS|NBEAL2|NBEL2_HUMAN|Neurobeachin-like protein 2|UNQ253/PRO290
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZNJ1
- Q6ZNJ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together