Order Entry
Canada
ContactUsLinkComponent
Anti-DTX1 Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-DTX1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-DTX1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77832-439
  • 77832-438
  • Antigen symbol:
  • DTX1
  • DTX1
  • Antigen name:
  • E3 ubiquitin-protein ligase DTX1 protein (261-410AA)
  • E3 ubiquitin-protein ligase DTX1 protein (261-410AA)
  • Antigen synonyms:
  • Deltex 1| E3 ubiquitin ligase| Deltex| Deltex homolog 1 (Drosophila)| Deltex homolog 1| Deltex protein 1| Deltex-1| Deltex1| dtx1| DTX1_HUMAN| E3 ubiquitin protein ligase DTX1| E3 ubiquitin-protein ligase DTX1| FXI-T1| Fxit 1| Fxit1| hDTX1| hDx 1| mDTX1| Protein deltex 1| Protein deltex-1
  • Deltex 1| E3 ubiquitin ligase| Deltex| Deltex homolog 1 (Drosophila)| Deltex homolog 1| Deltex protein 1| Deltex-1| Deltex1| dtx1| DTX1_HUMAN| E3 ubiquitin protein ligase DTX1| E3 ubiquitin-protein ligase DTX1| FXI-T1| Fxit 1| Fxit1| hDTX1| hDx 1| mDTX1| Protein deltex 1| Protein deltex-1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q86Y01
  • Q86Y01
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together