Anti-CASZ1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CASZ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-377
- 77832-376
- Antigen symbol:
- CASZ1
- CASZ1
- Antigen name:
- Zinc finger protein castor homolog 1 protein (127-266AA)
- Zinc finger protein castor homolog 1 protein (127-266AA)
- Antigen synonyms:
- 2410019P08Rik| AV096627| BC040081| BOS_15820| Castor-related protein| CASZ1| CASZ1_HUMAN| CST| D4Ertd432e| dJ734G22.1| FLJ12223| FLJ20321| MGC155102| MGC31683| OTTMUSP00000011746| OTTMUSP00000011748| Putative survival-related protein| RP23-230I5.1| RP4-734G22.2| SRG| Survival related| Zinc finger protein 693| Zinc finger protein castor homolog 1| ZNF693
- 2410019P08Rik| AV096627| BC040081| BOS_15820| Castor-related protein| CASZ1| CASZ1_HUMAN| CST| D4Ertd432e| dJ734G22.1| FLJ12223| FLJ20321| MGC155102| MGC31683| OTTMUSP00000011746| OTTMUSP00000011748| Putative survival-related protein| RP23-230I5.1| RP4-734G22.2| SRG| Survival related| Zinc finger protein 693| Zinc finger protein castor homolog 1| ZNF693
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86V15
- Q86V15
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together