Anti-REPS1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: REPS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-009
- 77834-010
- Antigen symbol:
- REPS1
- REPS1
- Antigen name:
- RalBP1-associated Eps domain-containing protein 1 protein (105-230AA)
- RalBP1-associated Eps domain-containing protein 1 protein (105-230AA)
- Antigen synonyms:
- RALBP1|RalBP1 associated Eps domain containing protein 1|RalBP1 interacting protein 1|RalBP1-associated Eps domain-containing protein 1|RalBP1-interacting protein 1|REPS1_HUMAN
- RALBP1|RalBP1 associated Eps domain containing protein 1|RalBP1 interacting protein 1|RalBP1-associated Eps domain-containing protein 1|RalBP1-interacting protein 1|REPS1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96D71
- Q96D71
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together