Anti-RNASEH2A Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RNASEH2A
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-416
- 77797-415
- Antigen symbol:
- RNASEH2A
- RNASEH2A
- Antigen name:
- Ribonuclease H2 subunit A protein (201-299AA)
- Ribonuclease H2 subunit A protein (201-299AA)
- Antigen synonyms:
- RNASEH2A|RNASEHI|RNHIA|Ribonuclease H2 subunit A|RNase H2 subunit A|EC 3.1.26.4|Aicardi-Goutieres syndrome 4 protein|AGS4|RNase H(35)|Ribonuclease HI large subunit|RNase HI large subunit|Ribonuclease HI subunit A
- RNASEH2A|RNASEHI|RNHIA|Ribonuclease H2 subunit A|RNase H2 subunit A|EC 3.1.26.4|Aicardi-Goutieres syndrome 4 protein|AGS4|RNase H(35)|Ribonuclease HI large subunit|RNase HI large subunit|Ribonuclease HI subunit A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75792
- O75792
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together