Anti-fabD Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: fabD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Staphylococcus aureus
Specifications
- Catalog No:
- 77826-902
- 77826-901
- Antigen symbol:
- fabD
- fabD
- Antigen name:
- Recombinant Staphylococcus aureus Malonyl CoA-acyl carrier protein transacylase protein (1-308AA)
- Recombinant Staphylococcus aureus Malonyl CoA-acyl carrier protein transacylase protein (1-308AA)
- Antigen synonyms:
- fabD|SACOL1244Malonyl CoA-acyl carrier protein transacylase|MCT|EC 2.3.1.39
- fabD|SACOL1244Malonyl CoA-acyl carrier protein transacylase|MCT|EC 2.3.1.39
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Staphylococcus aureus
- Staphylococcus aureus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5HGK3
- Q5HGK3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together