Anti-STX1A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:15 to 1:50
Type:
Antigen: STX1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Specifications
- Catalog No:
- 77813-218
- 77813-217
- Antigen symbol:
- STX1A
- STX1A
- Antigen name:
- Synthetic peptide of STX1A
- Synthetic peptide of STX1A
- Antigen synonyms:
- HPC 1|Neuron specific antigen HPC1|Neuron-specific antigen HPC-1|OTTHUMP00000174615|OTTHUMP00000174616|OTTHUMP00000174617|OTTHUMP00000174618|P35-1|STX1|STX1A|STX1A_HUMAN|SYN1A|Syntaxin 1A (brain)|Syntaxin 1A brain|Syntaxin-1A
- HPC 1|Neuron specific antigen HPC1|Neuron-specific antigen HPC-1|OTTHUMP00000174615|OTTHUMP00000174616|OTTHUMP00000174617|OTTHUMP00000174618|P35-1|STX1|STX1A|STX1A_HUMAN|SYN1A|Syntaxin 1A (brain)|Syntaxin 1A brain|Syntaxin-1A
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16623
- Q16623
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together