Anti-SULT1B1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77801-554
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Sulfotransferase family cytosolic 1B member 1 protein (1-271AA)
- Antigen symbol:SULT1B1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O43704
- Antigen synonyms:Dopa/tyrosine sulfotransferase| MGC13356| ST1B1| ST1B1_HUMAN| ST1B2| Sulfotransferase 1B1| Sulfotransferase 1B2| Sulfotransferase family 1B member 1| Sulfotransferase family cytosolic 1B member 1| SULT1B1| SULT1B2| Thyroid hormone sulfotransferase
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: SULT1B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse