Anti-S100A10 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: S100A10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77798-446
- 77798-447
- Antigen symbol:
- S100A10
- S100A10
- Antigen name:
- Protein S100-A10 protein (2-97AA)
- Protein S100-A10 protein (2-97AA)
- Antigen synonyms:
- 42C|AA409961|AL024248|Annexin II ligand|calpactin I|light polypeptide|Annexin II tetramer (AIIt) p11 subunit|Annexin II|light chain|ANX2L|ANX2LG|Ca[1]|CAL12|CAL1L|Calpactin I light chain|p11 subunit|Calpactin-1 light chain|Cellular ligand of annexin II|CLP11|GP11|MGC111133|Nerve growth factor-induced protein 42C|OTTHUMP00000015269|OTTHUMP00000015270|p10|p10 protein|p11|Protein S100 A10|Protein S100-A10|S100 calcium binding protein A10 (annexin II ligand|light polypeptide (p11))|S100 calcium binding protein A10 (calpactin)|S100 calcium binding protein A10|S100 calcium-binding protein A10|S100a10|S10AA_HUMAN
- 42C|AA409961|AL024248|Annexin II ligand|calpactin I|light polypeptide|Annexin II tetramer (AIIt) p11 subunit|Annexin II|light chain|ANX2L|ANX2LG|Ca[1]|CAL12|CAL1L|Calpactin I light chain|p11 subunit|Calpactin-1 light chain|Cellular ligand of annexin II|CLP11|GP11|MGC111133|Nerve growth factor-induced protein 42C|OTTHUMP00000015269|OTTHUMP00000015270|p10|p10 protein|p11|Protein S100 A10|Protein S100-A10|S100 calcium binding protein A10 (annexin II ligand|light polypeptide (p11))|S100 calcium binding protein A10 (calpactin)|S100 calcium binding protein A10|S100 calcium-binding protein A10|S100a10|S10AA_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P60903
- P60903
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together