Anti-RANBP9 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
About this item
Type:
Antigen: RANBP9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: RANBP9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications for Product Family
Specifications
- Catalog No:
- 77833-250
- 77833-251
- Antigen symbol:
- RANBP9
- RANBP9
- Antigen name:
- Ran-binding protein 9 protein (158-176AA)
- Ran-binding protein 9 protein (158-176AA)
- Antigen synonyms:
- B cell antigen receptor Ig beta associated protein 1|BPM 90|BPM L|BPM-L|BPM90|BPML|IBAP 1|Imp 9|Importin 9|Novel centrosomal protein RanBPM|RAN binding protein 9|Ran binding protein centrosomal|Ran Binding Protein in the Microtubule organizing center|Ran binding protein M|Ran BP9|Ran-binding protein 9|Ran-binding protein M|RANB9_HUMAN|RanBP 7|RANBP 9|RanBP7|RanBP9|RanBPM
- B cell antigen receptor Ig beta associated protein 1|BPM 90|BPM L|BPM-L|BPM90|BPML|IBAP 1|Imp 9|Importin 9|Novel centrosomal protein RanBPM|RAN binding protein 9|Ran binding protein centrosomal|Ran Binding Protein in the Microtubule organizing center|Ran binding protein M|Ran BP9|Ran-binding protein 9|Ran-binding protein M|RANB9_HUMAN|RanBP 7|RANBP 9|RanBP7|RanBP9|RanBPM
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96S59
- Q96S59
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Product Family Options
Product Information
- SizePackagingAvailabilityPrice
- Supplier Number: A65532-100ULItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77833-250Antigen symbolRANBP9Antigen nameRan-binding protein 9 protein (158-176AA)Antigen synonymsB cell antigen receptor Ig beta associated protein 1|BPM 90|BPM L|BPM-L|BPM90|BPML|IBAP 1|Imp 9|Importin 9|Novel centrosomal protein RanBPM|RAN binding protein 9|Ran binding protein centrosomal|Ran Binding Protein in the Microtubule organizing center|Ran binding protein M|Ran BP9|Ran-binding protein 9|Ran-binding protein M|RANB9_HUMAN|RanBP 7|RANBP 9|RanBP7|RanBP9|RanBPMClonalityPolyclonalConjugationBiotinReactivityHumanHostRabbitGene IDQ96S59IsotypeIgGELISAYesFormLiquidPurificationAntigen affinity purifiedStorage buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.Storage temperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping temperatureShipped on Blue ice - Supplier Number: A65532-50ULItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77833-251Antigen symbolRANBP9Antigen nameRan-binding protein 9 protein (158-176AA)Antigen synonymsB cell antigen receptor Ig beta associated protein 1|BPM 90|BPM L|BPM-L|BPM90|BPML|IBAP 1|Imp 9|Importin 9|Novel centrosomal protein RanBPM|RAN binding protein 9|Ran binding protein centrosomal|Ran Binding Protein in the Microtubule organizing center|Ran binding protein M|Ran BP9|Ran-binding protein 9|Ran-binding protein M|RANB9_HUMAN|RanBP 7|RANBP 9|RanBP7|RanBP9|RanBPMClonalityPolyclonalConjugationBiotinReactivityHumanHostRabbitGene IDQ96S59IsotypeIgGELISAYesFormLiquidPurificationAntigen affinity purifiedStorage buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.Storage temperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping temperatureShipped on Blue ice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



