Anti-TMC2 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: TMC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-574
- 77833-575
- Antigen symbol:
- TMC2
- TMC2
- Antigen name:
- Transmembrane channel-like protein 2 protein (6-206AA)
- Transmembrane channel-like protein 2 protein (6-206AA)
- Antigen synonyms:
- TMC2|C20orf145|UNQ907/PRO1928|Transmembrane channel-like protein 2|Transmembrane cochlear-expressed protein 2
- TMC2|C20orf145|UNQ907/PRO1928|Transmembrane channel-like protein 2|Transmembrane cochlear-expressed protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDI7
- Q8TDI7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together