Anti-SENP6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SENP6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-035
- 77837-036
- Antigen symbol:
- SENP6
- SENP6
- Antigen name:
- Sentrin-specific protease 6 protein (1-270AA)
- Sentrin-specific protease 6 protein (1-270AA)
- Antigen synonyms:
- 2810017C20Rik|FLJ11355|FLJ11887|KIAA0389|KIAA0797|SENP 6|Senp6|SENP6_HUMAN|Sentrin specific protease 6|Sentrin-specific protease 6|Sentrin/SUMO specific protease SENP6|Sentrin/SUMO-specific protease SENP6|SSP 1|SSP1|SUMO 1 specific protease 1|SUMO 1 specific protease|SUMO-1-specific protease 1|SUMO1 specific protease 1|SUMO1 specific protease|SUMO1/sentrin specific peptidase 6|SUSP 1|SUSP1
- 2810017C20Rik|FLJ11355|FLJ11887|KIAA0389|KIAA0797|SENP 6|Senp6|SENP6_HUMAN|Sentrin specific protease 6|Sentrin-specific protease 6|Sentrin/SUMO specific protease SENP6|Sentrin/SUMO-specific protease SENP6|SSP 1|SSP1|SUMO 1 specific protease 1|SUMO 1 specific protease|SUMO-1-specific protease 1|SUMO1 specific protease 1|SUMO1 specific protease|SUMO1/sentrin specific peptidase 6|SUSP 1|SUSP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZR1
- Q9GZR1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together