Anti-ST14 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: ST14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77826-648
- 77826-649
- Antigen symbol:
- ST14
- ST14
- Antigen name:
- Fusion protein of ST14
- Fusion protein of ST14
- Antigen synonyms:
- Epithin| HAI| Matriptase| Membrane type serine protease 1| Membrane type serine protease| Membrane-type serine protease 1| MT SP1| MT-SP1| MTSP 1| MTSP1| Prostamin| PRSS 14| PRSS14| Serine protease 14| Serine protease TADG 15| Serine protease TADG-15| Serine protease TADG15| SNC 19| SNC19| ST 14| ST14| ST14_HUMAN| Suppression of tumorigenicity 14 (colon carcinoma matriptase epithin)| Suppression of tumorigenicity 14 (colon carcinoma)| Suppression of tumorigenicity 14| Suppressor of tumorigenicity 14 protein| Suppressor of tumorigenicity protein 14| TADG 15| TADG15| TMPRSS14| Tumor associated differentially expressed gene 15 protein| Tumor-associated differentially-expressed gene 15 protein
- Epithin| HAI| Matriptase| Membrane type serine protease 1| Membrane type serine protease| Membrane-type serine protease 1| MT SP1| MT-SP1| MTSP 1| MTSP1| Prostamin| PRSS 14| PRSS14| Serine protease 14| Serine protease TADG 15| Serine protease TADG-15| Serine protease TADG15| SNC 19| SNC19| ST 14| ST14| ST14_HUMAN| Suppression of tumorigenicity 14 (colon carcinoma matriptase epithin)| Suppression of tumorigenicity 14 (colon carcinoma)| Suppression of tumorigenicity 14| Suppressor of tumorigenicity 14 protein| Suppressor of tumorigenicity protein 14| TADG 15| TADG15| TMPRSS14| Tumor associated differentially expressed gene 15 protein| Tumor-associated differentially-expressed gene 15 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5Y6
- Q9Y5Y6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together