Anti-CNNM4 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: CNNM4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77831-416
- 77831-417
- Antigen symbol:
- CNNM4
- CNNM4
- Antigen name:
- Metal transporter CNNM4 protein (589-691AA)
- Metal transporter CNNM4 protein (589-691AA)
- Antigen synonyms:
- 430430O18Rik|ACDP4|Ancient conserved domain protein 4|Ancient conserved domain-containing protein 4|cnnm4|CNNM4_HUMAN|Cyclin and CBS domain divalent metal cation transport mediator 4|Cyclin M4|Cyclin-M4|FLJ42791|KIAA1592|Metal transporter CNNM4
- 430430O18Rik|ACDP4|Ancient conserved domain protein 4|Ancient conserved domain-containing protein 4|cnnm4|CNNM4_HUMAN|Cyclin and CBS domain divalent metal cation transport mediator 4|Cyclin M4|Cyclin-M4|FLJ42791|KIAA1592|Metal transporter CNNM4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P4Q7
- Q6P4Q7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together