Anti-SLC25A25 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC25A25
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-116
- 77831-117
- Antigen symbol:
- SLC25A25
- SLC25A25
- Antigen name:
- Calcium-binding mitochondrial carrier protein SCaMC-2 protein (1-189AA)
- Calcium-binding mitochondrial carrier protein SCaMC-2 protein (1-189AA)
- Antigen synonyms:
- Calcium-binding mitochondrial carrier protein SCaMC-2|KIAA1896|MCSC|MGC105138|MGC119514|MGC119515|MGC119516|MGC119517|Mitochondrial ATP Mg/Pi carrier|Mitochondrial ATP-Mg/Pi carrier protein 3|Mitochondrial Ca(2+)-dependent solute carrier protein 3|Mitochondrial Ca2+ dependent solute carrier|Mitochondrial calcium-dependent solute carrier|PCSCL|RP11 395P17.4|SCAMC 2|SCAMC2|SCMC2_HUMAN|Short calcium binding mitochondrial carrier 2|SLC25A25|Small calcium binding mitochondrial carrier 2|Small calcium-binding mitochondrial carrier protein 2|Solute carrier family 25 (mitochondrial carrier phosphate carrier) member 25|Solute carrier family 25 member 25|solute carrier family 25, member 25, isoform a
- Calcium-binding mitochondrial carrier protein SCaMC-2|KIAA1896|MCSC|MGC105138|MGC119514|MGC119515|MGC119516|MGC119517|Mitochondrial ATP Mg/Pi carrier|Mitochondrial ATP-Mg/Pi carrier protein 3|Mitochondrial Ca(2+)-dependent solute carrier protein 3|Mitochondrial Ca2+ dependent solute carrier|Mitochondrial calcium-dependent solute carrier|PCSCL|RP11 395P17.4|SCAMC 2|SCAMC2|SCMC2_HUMAN|Short calcium binding mitochondrial carrier 2|SLC25A25|Small calcium binding mitochondrial carrier 2|Small calcium-binding mitochondrial carrier protein 2|Solute carrier family 25 (mitochondrial carrier phosphate carrier) member 25|Solute carrier family 25 member 25|solute carrier family 25, member 25, isoform a
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6KCM7
- Q6KCM7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together