Anti-ITSN2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77838-943
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Intersectin-2 protein (940-1209AA)
- Antigen symbol:ITSN2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NZM3
- Antigen synonyms:ITSN2|KIAA1256|SH3D1B|SWAP|Intersectin-2|SH3 domain-containing protein 1B|SH3P18|SH3P18-like WASP-associated protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:50 to 1:200
Type:
Antigen: ITSN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human