Anti-CNNM1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CNNM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-788
- 77839-787
- Antigen symbol:
- CNNM1
- CNNM1
- Antigen name:
- Metal transporter CNNM1 protein (287-586AA)
- Metal transporter CNNM1 protein (287-586AA)
- Antigen synonyms:
- ACDP1|Ancient conserved domain protein 1|Ancient conserved domain-containing protein 1|CNNM1|CNNM1_HUMAN|Cyclin-M1|FLJ31632|Metal transporter CNNM1|OTTHUMP00000020253
- ACDP1|Ancient conserved domain protein 1|Ancient conserved domain-containing protein 1|CNNM1|CNNM1_HUMAN|Cyclin-M1|FLJ31632|Metal transporter CNNM1|OTTHUMP00000020253
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NRU3
- Q9NRU3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together