Anti-PDE2A Rabbit Recombinant Polyclonal Antibody
Catalog # 77793-554
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:cGMP-dependent 3\\\',5\\\'-cyclic phosphodiesterase protein (15-226AA)
- Antigen symbol:PDE2A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O00408
- Antigen synonyms:5''-cyclic phosphodiesterase|cGMP dependent 3'5' cyclic phosphodiesterase|cGMP stimulated phosphodiesterase 1|cGMP stimulated phosphodiesterase 4|cGMP-dependent 3''|cGMP-stimulated phosphodiesterase 2|CGS PDE|CGS-PDE|cGSPDE|Cyclic GMP stimulated phosphodiesterase|Cyclic GMP-stimulated phosphodiesterase|Human cGMP stimulated 3'5' cyclic nucleotide phosphodiesterase|PDE 2A|PDE 2A1|PDE 2A4|PDE2A|PDE2A_HUMAN|PDE2A1|PDE2A4|phosphodiesterase 2A|Phosphodiesterase 2A cGMP stimulated
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PDE2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human