Anti-PPA1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: PPA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-616
- 77822-617
- Antigen symbol:
- PPA1
- PPA1
- Antigen name:
- Inorganic pyrophosphatase protein (59-173AA)
- Inorganic pyrophosphatase protein (59-173AA)
- Antigen synonyms:
- cytosolic inorganic pyrophosphatase|diphosphate phosphohydrolase|inorganic diphosphatase|inorganic pyrophosphatase 1|Inorganic pyrophosphatase|IOPPP|IPYR_HUMAN|PP|PP1|PPA1|PPase|Pyp|pyrophosphatase, inorganic, 1|Pyrophosphate phospho hydrolase|Pyrophosphate phospho-hydrolase|SID6-8061
- cytosolic inorganic pyrophosphatase|diphosphate phosphohydrolase|inorganic diphosphatase|inorganic pyrophosphatase 1|Inorganic pyrophosphatase|IOPPP|IPYR_HUMAN|PP|PP1|PPA1|PPase|Pyp|pyrophosphatase, inorganic, 1|Pyrophosphate phospho hydrolase|Pyrophosphate phospho-hydrolase|SID6-8061
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15181
- Q15181
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together