Anti-RBBP7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RBBP7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-600
- 77824-599
- Antigen symbol:
- RBBP7
- RBBP7
- Antigen name:
- Histone-binding protein RBBP7 protein (1-425AA)
- Histone-binding protein RBBP7 protein (1-425AA)
- Antigen synonyms:
- G1/S transition control protein binding protein RbAp46|Histone acetyltransferase type B subunit 2|Histone binding protein |Histone-binding protein |MGC138867|MGC138868|Nucleosome remodeling factor subunit RBAP46|Nucleosome-remodeling factor subunit RBAP46|RBAP46|RBBP 7|RBBP-7||_HUMAN|Retinoblastoma binding protein 7|Retinoblastoma binding protein p46|Retinoblastoma-binding protein 7|Retinoblastoma-binding protein p46|Retinoblastoma-binding protein RbAp46
- G1/S transition control protein binding protein RbAp46|Histone acetyltransferase type B subunit 2|Histone binding protein |Histone-binding protein |MGC138867|MGC138868|Nucleosome remodeling factor subunit RBAP46|Nucleosome-remodeling factor subunit RBAP46|RBAP46|RBBP 7|RBBP-7||_HUMAN|Retinoblastoma binding protein 7|Retinoblastoma binding protein p46|Retinoblastoma-binding protein 7|Retinoblastoma-binding protein p46|Retinoblastoma-binding protein RbAp46
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16576
- Q16576
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together