Anti-CAPRIN1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: CAPRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77814-003
- 77814-004
- Antigen symbol:
- CAPRIN1
- CAPRIN1
- Antigen name:
- Fusion protein of CAPRIN1
- Fusion protein of CAPRIN1
- Antigen synonyms:
- activation/proliferation-associated protein 1|CAPR1_HUMAN|Caprin-1|CAPRIN1|Cell cycle-associated protein 1|Cytoplasmic activation- and proliferation-associated protein 1|cytoplasmic activation/proliferation-associated protein-1|GPI-anchored membrane protein 1|GPI-anchored protein p137|GPI-p137|GPIAP1|GPIP137|M11S1|Membrane component chromosome 11 surface marker 1|p137GPI|RNA granule protein 105|RNG105
- activation/proliferation-associated protein 1|CAPR1_HUMAN|Caprin-1|CAPRIN1|Cell cycle-associated protein 1|Cytoplasmic activation- and proliferation-associated protein 1|cytoplasmic activation/proliferation-associated protein-1|GPI-anchored membrane protein 1|GPI-anchored protein p137|GPI-p137|GPIAP1|GPIP137|M11S1|Membrane component chromosome 11 surface marker 1|p137GPI|RNA granule protein 105|RNG105
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14444
- Q14444
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together