Anti-RPL10A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RPL10A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-511
- 77808-510
- Antigen symbol:
- RPL10A
- RPL10A
- Antigen name:
- 60S ribosomal protein L10a protein (40-80AA)
- 60S ribosomal protein L10a protein (40-80AA)
- Antigen synonyms:
- 60S ribosomal protein L10a|CSA 19|CSA-19|CSA19|L10A|NEDD 6|NEDD-6|NEDD6|neural precursor cell expressed developmentally down regulated protein 6|Neural precursor cell expressed developmentally down-regulated protein 6|Protein NEDD6|Ribosomal protein L10a|RL10A_HUMAN|rpl10a
- 60S ribosomal protein L10a|CSA 19|CSA-19|CSA19|L10A|NEDD 6|NEDD-6|NEDD6|neural precursor cell expressed developmentally down regulated protein 6|Neural precursor cell expressed developmentally down-regulated protein 6|Protein NEDD6|Ribosomal protein L10a|RL10A_HUMAN|rpl10a
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62906
- P62906
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together