Anti-dsbA Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: dsbA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pseudomonas syringae pv. tomato
- Catalog No:
- 77819-493
- 77819-494
- Antigen symbol:
- dsbA
- dsbA
- Antigen name:
- Recombinant Pseudomonas syringae pv. tomato Thiol:disulfide interchange protein DsbA protein (23-214AA)
- Recombinant Pseudomonas syringae pv. tomato Thiol:disulfide interchange protein DsbA protein (23-214AA)
- Antigen synonyms:
- dsbA|PSPTO_0341Thiol:disulfide interchange protein DsbA
- dsbA|PSPTO_0341Thiol:disulfide interchange protein DsbA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Pseudomonas syringae pv. tomato
- Pseudomonas syringae pv. tomato
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O52376
- O52376
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together