Anti-ARL15 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ARL15
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-093
- 77841-094
- Antigen symbol:
- ARL15
- ARL15
- Antigen name:
- ADP-ribosylation factor-like protein 15 protein (1-204AA)
- ADP-ribosylation factor-like protein 15 protein (1-204AA)
- Antigen synonyms:
- A430036I03| ADP ribosylation factor like 15 | ADP ribosylation factor like GTPase 15| ADP-ribosylation factor-like protein 15| ADP-ribosylation factor-related protein 2| ARF-related protein 2| ARFRP2| ARL15| ARL15_HUMAN| C230032K13Rik| FLJ20051| rCG_44824
- A430036I03| ADP ribosylation factor like 15 | ADP ribosylation factor like GTPase 15| ADP-ribosylation factor-like protein 15| ADP-ribosylation factor-related protein 2| ARF-related protein 2| ARFRP2| ARL15| ARL15_HUMAN| C230032K13Rik| FLJ20051| rCG_44824
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NXU5
- Q9NXU5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together