Anti-CDON Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: CDON
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77826-655
- 77826-656
- Antigen symbol:
- CDON
- CDON
- Antigen name:
- Cell adhesion molecule-related/down-regulated by oncogenes protein (1095-1211AA)
- Cell adhesion molecule-related/down-regulated by oncogenes protein (1095-1211AA)
- Antigen synonyms:
- CDO|Cdon|CDON_HUMAN|Cell adhesion molecule-related/down-regulated by oncogenes
- CDO|Cdon|CDON_HUMAN|Cell adhesion molecule-related/down-regulated by oncogenes
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q4KMG0
- Q4KMG0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together