Anti-LIN9 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LIN9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-019
- 77828-018
- Antigen symbol:
- LIN9
- LIN9
- Antigen name:
- Protein lin-9 homolog protein (5-103AA)
- Protein lin-9 homolog protein (5-103AA)
- Antigen synonyms:
- BARA|Beta subunit-associated regulator of apoptosis|Beta-subunit associated regulator of apoptosis|hLin-9|huLin-9|lin-9|lin9|LIN9_HUMAN|pRB-associated protein|Protein lin-9 homolog|TGS|TUDOR gene similar protein|Type I interferon receptor beta chain-associated protein
- BARA|Beta subunit-associated regulator of apoptosis|Beta-subunit associated regulator of apoptosis|hLin-9|huLin-9|lin-9|lin9|LIN9_HUMAN|pRB-associated protein|Protein lin-9 homolog|TGS|TUDOR gene similar protein|Type I interferon receptor beta chain-associated protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5TKA1
- Q5TKA1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together