Anti-POLR3E Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: POLR3E
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-531
- 77837-530
- Antigen symbol:
- POLR3E
- POLR3E
- Antigen name:
- DNA-directed RNA polymerase III subunit RPC5 protein (1-260AA)
- DNA-directed RNA polymerase III subunit RPC5 protein (1-260AA)
- Antigen synonyms:
- POLR3E|KIAA1452|DNA-directed RNA polymerase III subunit RPC5|RNA polymerase III subunit C5|DNA-directed RNA polymerase III 80 kDa polypeptide
- POLR3E|KIAA1452|DNA-directed RNA polymerase III subunit RPC5|RNA polymerase III subunit C5|DNA-directed RNA polymerase III 80 kDa polypeptide
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVU0
- Q9NVU0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together