Anti-SGCE Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SGCE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-404
- 77799-403
- Antigen symbol:
- SGCE
- SGCE
- Antigen name:
- Epsilon-sarcoglycan protein (8-126AA)
- Epsilon-sarcoglycan protein (8-126AA)
- Antigen synonyms:
- dystonia 11, myoclonic|DYT11|Epsilon sarcoglycan|Epsilon SG|Epsilon-sarcoglycan|Epsilon-SG|ESG|sarcoglycan, epsilon|sgcE|SGCE_HUMAN
- dystonia 11, myoclonic|DYT11|Epsilon sarcoglycan|Epsilon SG|Epsilon-sarcoglycan|Epsilon-SG|ESG|sarcoglycan, epsilon|sgcE|SGCE_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43556
- O43556
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together