Anti-SPANXC Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SPANXC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77839-983
- 77839-982
- Antigen symbol:
- SPANXC
- SPANXC
- Antigen name:
- Sperm protein associated with the nucleus on the X chromosome C protein (1-97AA)
- Sperm protein associated with the nucleus on the X chromosome C protein (1-97AA)
- Antigen synonyms:
- Cancer-testis-associated protein CTp11|Cancer/testis antigen 11.3|Cancer/testis-associated protein CTp11|CT11.3|CTP11|Nuclear associated protein SPAN Xc|Nuclear-associated protein SPAN-Xc|SPANX family member C|SPANX family|member C|member C1|SPANX-C|SPANXC|Sperm protein associated with the nucleus on the X chromosome C|SPNXC_HUMAN
- Cancer-testis-associated protein CTp11|Cancer/testis antigen 11.3|Cancer/testis-associated protein CTp11|CT11.3|CTP11|Nuclear associated protein SPAN Xc|Nuclear-associated protein SPAN-Xc|SPANX family member C|SPANX family|member C|member C1|SPANX-C|SPANXC|Sperm protein associated with the nucleus on the X chromosome C|SPNXC_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NY87
- Q9NY87
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together