Anti-ATG16L1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ATG16L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-080
- 77829-079
- Antigen symbol:
- ATG16L1
- ATG16L1
- Antigen name:
- Autophagy-related protein 16-1 protein (1-607AA)
- Autophagy-related protein 16-1 protein (1-607AA)
- Antigen synonyms:
- A16L1_HUMAN|APG16 like 1|APG16-like 1|APG16L|APG16L beta|ATG16 autophagy related 16 like 1|ATG16 autophagy related 16-like 1 (S. cerevisiae)|ATG16A|ATG16L|Atg16l1|Autophagy related protein 16 1|Autophagy-related protein 16-1|FLJ00045|FLJ10035|FLJ10828|FLJ22677|IBD10|OTTHUMP00000164391|OTTHUMP00000164393|OTTHUMP00000165876|OTTHUMP00000165877|WD repeat domain 30|WDR30
- A16L1_HUMAN|APG16 like 1|APG16-like 1|APG16L|APG16L beta|ATG16 autophagy related 16 like 1|ATG16 autophagy related 16-like 1 (S. cerevisiae)|ATG16A|ATG16L|Atg16l1|Autophagy related protein 16 1|Autophagy-related protein 16-1|FLJ00045|FLJ10035|FLJ10828|FLJ22677|IBD10|OTTHUMP00000164391|OTTHUMP00000164393|OTTHUMP00000165876|OTTHUMP00000165877|WD repeat domain 30|WDR30
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q676U5
- Q676U5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together