Anti-SERF1A Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SERF1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-077
- 77799-078
- Antigen symbol:
- SERF1A
- SERF1A
- Antigen name:
- Small EDRK-rich factor 1 protein (1-74AA)
- Small EDRK-rich factor 1 protein (1-74AA)
- Antigen synonyms:
- SERF1A|FAM2A|SERF1|SMAM1||SERF1B|FAM2B|Small EDRK-rich factor 1|Protein 4F5|h4F5|SMA modifier 1
- SERF1A|FAM2A|SERF1|SMAM1||SERF1B|FAM2B|Small EDRK-rich factor 1|Protein 4F5|h4F5|SMA modifier 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75920
- O75920
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together