Anti-TDP1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77802-185
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Tyrosyl-DNA phosphodiesterase 1 protein (1-260AA)
- Antigen symbol:TDP1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9NUW8
- Antigen synonyms:AI838772|AW493413|FLJ11090|MGC104252|MGC112732|RP24-311F12.2|SCAN1|TDP1|TYDP|TYDP1_HUMAN|Tyr-DNA phosphodiesterase 1|Tyrosyl-DNA phosphodiesterase 1
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:2000, IHC: 1:20 to 1:200, IP:1:200 to 1:2000
Type:
Antigen: TDP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat