Anti-SCD Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SCD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77798-724
- 77798-725
- Antigen symbol:
- SCD
- SCD
- Antigen name:
- Acyl-CoA desaturase protein (10-70AA)
- Acyl-CoA desaturase protein (10-70AA)
- Antigen synonyms:
- ACOD_HUMAN|Acyl-CoA desaturase|Delta(9)-desaturase|Delta-9 desaturase|Delta-9-Desaturase|FADS5|Fatty acid desaturase|PRO0998|SCD 1|SCD1|Stearoyl-CoA desaturase
- ACOD_HUMAN|Acyl-CoA desaturase|Delta(9)-desaturase|Delta-9 desaturase|Delta-9-Desaturase|FADS5|Fatty acid desaturase|PRO0998|SCD 1|SCD1|Stearoyl-CoA desaturase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00767
- O00767
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together