Order Entry
Canada
ContactUsLinkComponent
Anti-GJC2 Rabbit Polyclonal Antibody
Anti-GJC2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-GJC2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77809-810
  • 77809-809
  • Antigen symbol:
  • GJC2
  • GJC2
  • Antigen name:
  • Synthetic peptide of GJC2
  • Synthetic peptide of GJC2
  • Antigen synonyms:
  • B230382L12Rik|Connexin 46.6|Connexin 47|Connexin-46.6|Connexin-47|Connexin46.6|Connexin47|CX 46.6|Cx 47|Cx46.6|Cx47|CXG2_HUMAN|Gap junction alpha 12 protein|Gap junction alpha-12 protein|Gap junction gamma 2 protein|Gap junction gamma-2 protein|Gap junction protein alpha 12 47kDa|Gap junction protein gamma 2 47kDa |GAP JUNCTION PROTEIN|47-KD|gap junction protein|gamma 12|47kDa |gamma 2|47kDa|GJA 12|GJA12|GJC 2|Gjc2|HLD 2|HLD2|LMPH1C|MGC105119|MGC130506|OTTHUMP00000037763 |PMLDAR|SPG44
  • B230382L12Rik|Connexin 46.6|Connexin 47|Connexin-46.6|Connexin-47|Connexin46.6|Connexin47|CX 46.6|Cx 47|Cx46.6|Cx47|CXG2_HUMAN|Gap junction alpha 12 protein|Gap junction alpha-12 protein|Gap junction gamma 2 protein|Gap junction gamma-2 protein|Gap junction protein alpha 12 47kDa|Gap junction protein gamma 2 47kDa |GAP JUNCTION PROTEIN|47-KD|gap junction protein|gamma 12|47kDa |gamma 2|47kDa|GJA 12|GJA12|GJC 2|Gjc2|HLD 2|HLD2|LMPH1C|MGC105119|MGC130506|OTTHUMP00000037763 |PMLDAR|SPG44
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse, Rat
  • Human, Mouse, Rat
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q5T442
  • Q5T442
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together