Anti-PLD2 Rabbit Polyclonal Antibody
Catalog # 77806-210
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from PC-PLD2 around the phosphorylation site of Y169
- Antigen symbol:PLD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O14939
- Antigen synonyms:Choline phosphatase 2|EC 3.1.4.4|hPLD 2|hPLD2|mPLD2|Phosphatidylcholine hydrolyzing phospholipase D2|Phosphatidylcholine-hydrolyzing phospholipase D2|Phospholipase D2|PhospholipaseD2|PLD 2|PLD1C|Pld2|PLD2_HUMAN|Pldc|rPLD2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: PLD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat