Anti-GLDC Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: GLDC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77815-794
- 77815-793
- Antigen symbol:
- GLDC
- GLDC
- Antigen name:
- Glycine dehydrogenase (decarboxylating), mitochondrial protein (868-984AA)
- Glycine dehydrogenase (decarboxylating), mitochondrial protein (868-984AA)
- Antigen synonyms:
- GCE|GCSP|GCSP_HUMAN|GLDC|Glycine cleavage system P protein|glycine cleavage system protein P|Glycine decarboxylase|glycine decarboxylase P protein |Glycine dehydrogenase (decarboxylating) mitochondrial|Glycine dehydrogenase [decarboxylating]|mitochondrial|Gycine dehydrogenase (decarboxylating)|HYGN1|MGC138198|MGC138200|NKH
- GCE|GCSP|GCSP_HUMAN|GLDC|Glycine cleavage system P protein|glycine cleavage system protein P|Glycine decarboxylase|glycine decarboxylase P protein |Glycine dehydrogenase (decarboxylating) mitochondrial|Glycine dehydrogenase [decarboxylating]|mitochondrial|Gycine dehydrogenase (decarboxylating)|HYGN1|MGC138198|MGC138200|NKH
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P23378
- P23378
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together